missing translation for 'onlineSavingsMsg'
Learn More

Cinnamylideneacetophenone, 98+%

Catalog No. AAA1511414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1511414 25 g
AAA1511406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1511414 Supplier Thermo Scientific Chemicals Supplier No. A1511414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-57-3 | C17H14O | 234.298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-57-3
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.298
MDL Number MFCD00020692
InChI Key QONKLJMPKWQQFG-HPIZBCMHSA-N
Synonym cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
PubChem CID 1549519
IUPAC Name (2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
SMILES C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 100°C to 102°C
Quantity 25 g
Beilstein 1870052
Formula Weight 234.3
Percent Purity ≥98%
Chemical Name or Material Cinnamylideneacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-385-9
RTECSNumber RZ2477000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only