missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamyl Chloride 95.0+%, TCI America™
Supplier: TCI America C123525G
Specifications
| Cinnamyl Chloride | |
| Yellow | |
| C9H9Cl | |
| 25 g | |
| cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride | |
| C1=CC=C(C=C1)C=CCCl | |
| 152.62 | |
| 152.62 | |
| Liquid |
| 2687-12-9 | |
| 136°C | |
| MFCD00000986 | |
| 2927 | |
| IWTYTFSSTWXZFU-QPJJXVBHSA-N | |
| [(E)-3-chloroprop-1-enyl]benzene | |
| 639658 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 2687-12-9 | |
| 152.62 | |
| IWTYTFSSTWXZFU-QPJJXVBHSA-N | |
| 639658 | |
| C1=CC=C(C=C1)C=CCCl |
| C9H9Cl | |
| MFCD00000986 | |
| cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride | |
| [(E)-3-chloroprop-1-enyl]benzene |
Safety and Handling
EINECSNumber : (3)-2690
RTECSNumber : CZ0919905
TSCA : Yes