missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl chloride, 95% trans

Catalog No. p-93178
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181850050 5 g Glass bottle
AC181851000 100 g Glass bottle
AC181855000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC181850050 Supplier Thermo Scientific Chemicals Supplier No. 181850050

CAS: 2687-12-9 | C9H9Cl | 152.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2687-12-9
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000986
InChI Key IWTYTFSSTWXZFU-QPJJXVBHSA-N
Synonym cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride
PubChem CID 639658
IUPAC Name [(E)-3-chloroprop-1-enyl]benzene
SMILES C1=CC=C(C=C1)C=CCCl

Specifications

Melting Point 7.0°C
Color Brown-Yellow to Yellow
Density 1.0960g/mL
Boiling Point 120.0°C to 126.0°C (15.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2Cl
Refractive Index 1.5830 to 1.5850
Quantity 5 g
Beilstein 05, II, 372
Fieser 13,299
Specific Gravity 1.096
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, n,n-dmf,, acetone, formic acid, benzene, ethyl ether
Formula Weight 152.62
Percent Purity 95% trans
Physical Form Liquid
Chemical Name or Material Cinnamyl chloride, 95% trans
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Fatal if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 220-246-4

RUO – Research Use Only