missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl chloride, 95%, stab. with sodium carbonate

Catalog No. AAA1440818
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1440818 50 g
AAA1440830 250 g
AAA144080B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1440818 Supplier Thermo Scientific Chemicals Supplier No. A1440818

CAS: 2687-12-9 | C9H9Cl | 152.62 g/mol

Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C

Chemical Identifiers

CAS 2687-12-9

Specifications

Melting Point 7°C
Density 1.096
Boiling Point 86°C to 87°C (2 mmHg)
Flash Point 79°C (174°F)
Molecular Formula C9H9Cl
Refractive Index 1.5845
Quantity 50 g
MDL Number MFCD00000986
UN Number UN2927
Beilstein 741974
Synonym cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride
Solubility Information Soluble in water (0.2g/L at 20°C).
InChI Key IWTYTFSSTWXZFU-QPJJXVBHSA-N
SMILES C1=CC=C(C=C1)C=CCCl
IUPAC Name [(E)-3-chloroprop-1-enyl]benzene
Molecular Weight (g/mol) 152.62
PubChem CID 639658
Formula Weight 152.62
Percent Purity 95%
Chemical Name or Material Cinnamyl chloride, stabilized with sodium carbonate
Show More Show Less
Hazard Category H227-H302-H314-H317-H330-H334-H335
Hazard Statement GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 220-246-4
RTECSNumber CZ0919905
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only