missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl Butyrate 97.0+%, TCI America™

Catalog No. C243825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C243825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C243825G Supplier TCI America Supplier No. C243825G
Only null left

Chemical Identifiers

CAS 103-61-7
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00027135
InChI Key YZYPQKZWNXANRB-UXBLZVDNSA-N
Synonym Butyric Acid Cinnamyl Ester
PubChem CID 5355254
IUPAC Name [(E)-3-phenylprop-2-enyl] butanoate
SMILES CCCC(=O)OCC=CC1=CC=CC=C1

Specifications

Color Colorless
Quantity 25 g
Formula Weight 204.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cinnamyl Butyrate
EINECSNumber (3)-1043
RTECSNumber ES8815000
TSCA Yes