missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl Bromide 98.0+%, TCI America™

Catalog No. C110925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C110925G Supplier TCI America Supplier No. C110925G

Chemical Identifiers

CAS 4392-24-9
Molecular Formula C9H9Br
Molecular Weight (g/mol) 197.075
MDL Number MFCD00000245
InChI Key RUROFEVDCUGKHD-QPJJXVBHSA-N
Synonym cinnamyl bromide, 3-bromoprop-1-en-1-yl benzene, e-3-bromoprop-1-en-1-yl benzene, 3-bromo-1-phenyl-1-propene, 3-bromo-1-phenyl propene-1, e-3-bromoprop-1-enyl benzene, 3-bromo-propenyl-benzene, e-cinnamyl bromide, trans-cinnamyl bromide
PubChem CID 5357478
IUPAC Name [(E)-3-bromoprop-1-enyl]benzene
SMILES C1=CC=C(C=C1)C=CCBr

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 g
UN Number 3261
Formula Weight 197.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cinnamyl Bromide
TSCA No
Recommended Storage Refrigerator