missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl Acetate 97.0+%, TCI America™

Catalog No. A090125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A090125ML 25 mL
A0901500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A090125ML Supplier TCI America Supplier No. A090125ML
Only null left

Chemical Identifiers

CAS 103-54-8
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00008722
InChI Key WJSDHUCWMSHDCR-VMPITWQZSA-N
Synonym cinnamyl acetate, 3-phenyl-2-propenyl acetate, 3-phenylallyl acetate, cinnamyl alcohol, acetate, acetic acid, cinnamyl ester, trans-cinnamyl acetate, 3-phenyl-2-propen-1-yl acetate, gamma-phenylallyl acetate, 1-acetoxy-3-phenyl-2-propene, 3-phenyl-2-propen-1-ol acetate
PubChem CID 5282110
IUPAC Name [(E)-3-phenylprop-2-enyl] acetate
SMILES CC(=O)OCC=CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 265°C
Quantity 25 mL
Formula Weight 176.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cinnamyl Acetate
EINECSNumber (3)-1043
RTECSNumber GE2275000
TSCA Yes