missing translation for 'onlineSavingsMsg'
Learn More

Cinnamoyl Chloride 97.0+%, TCI America™

$59.69 - $59.69

Chemical Identifiers

CAS 17082-09-6
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.60
MDL Number MFCD00000732
InChI Key WOGITNXCNOTRLK-VOTSOKGWSA-N
Synonym cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
PubChem CID 5354261
IUPAC Name (2E)-3-phenylprop-2-enoyl chloride
SMILES ClC(=O)\C=C\C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P013325G
View Documents
TCI America
P013325G
25 g
Each for $59.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17082-09-6
166.60
WOGITNXCNOTRLK-VOTSOKGWSA-N
5354261
ClC(=O)\C=C\C1=CC=CC=C1
C9H7ClO
MFCD00000732
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
(2E)-3-phenylprop-2-enoyl chloride

Specifications

17082-09-6
C9H7ClO
25 g
cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
ClC(=O)\C=C\C1=CC=CC=C1
166.60
166.60
Crystalline Lumps
White-Yellow
MFCD00000732
3261
WOGITNXCNOTRLK-VOTSOKGWSA-N
(2E)-3-phenylprop-2-enoyl chloride
5354261
≥97.0% (T)
Cinnamoyl Chloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1720

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'