missing translation for 'onlineSavingsMsg'
Learn More

Cinnamonitrile, 97%, predominantly trans, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 110380250

 View more versions of this product

Catalog No. AC110380250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cinnamonitrile
1885-38-7
254°C to 255°C
Authentic
Glass bottle
1.6000 to 1.602
MFCD00001930
09, 589
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
(E)-3-phenylprop-2-enenitrile
1550846
97%
97%
1.0200g/mL
126°C
10% max. (cis-Cinnamonitrile) (GC)
C9H7N
C6H5CH=CHCN
25g
1.02
Solubility in water: insoluble
C1=CC=C(C=C1)C=CC#N
129.16
129.16

Chemical Identifiers

1885-38-7
129.16
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
1550846
C1=CC=C(C=C1)C=CC#N
C9H7N
MFCD00001930
cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
(E)-3-phenylprop-2-enenitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 217-552-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only