missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamaldoxime, (E)+(Z), 98%
CAS: 13372-81-1 | C9H9NO | 147.18 g/mol
$67.08 - $190.66
Chemical Identifiers
| CAS | 13372-81-1 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.18 |
| MDL Number | MFCD00019969 |
| InChI Key | RUQDOYIAKHIMAN-DAAQNPAKSA-N |
| Synonym | cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z |
| PubChem CID | 9561350 |
| IUPAC Name | (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine |
| SMILES | O\N=C\C=C\C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0502606
|
Thermo Scientific Chemicals
L0502606 |
5 g |
Each for $67.08
|
|
|||||
|
AAL0502614
|
Thermo Scientific Chemicals
L0502614 |
25 g |
Each for $190.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13372-81-1 | |
| 147.18 | |
| RUQDOYIAKHIMAN-DAAQNPAKSA-N | |
| 9561350 | |
| O\N=C\C=C\C1=CC=CC=C1 |
| C9H9NO | |
| MFCD00019969 | |
| cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z | |
| (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine |
Specifications
| 13372-81-1 | |
| C9H9NO | |
| 5 g | |
| cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z | |
| O\N=C\C=C\C1=CC=CC=C1 | |
| 147.18 | |
| 147.18 | |
| Cinnamaldoxime, (E)+(Z) |
| 117°C to 122°C | |
| MFCD00019969 | |
| 3117555 | |
| RUQDOYIAKHIMAN-DAAQNPAKSA-N | |
| (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine | |
| 9561350 | |
| 98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 236-449-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only