missing translation for 'onlineSavingsMsg'
Learn More

Cinnamaldoxime, (E)+(Z), 98%

Catalog No. AAL0502606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0502606 5 g
AAL0502614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0502606 Supplier Thermo Scientific Chemicals Supplier No. L0502606

CAS: 13372-81-1 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13372-81-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00019969
InChI Key RUQDOYIAKHIMAN-DAAQNPAKSA-N
Synonym cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z
PubChem CID 9561350
IUPAC Name (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine
SMILES O\N=C\C=C\C1=CC=CC=C1

Specifications

Melting Point 117°C to 122°C
Quantity 5 g
Beilstein 3117555
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material Cinnamaldoxime, (E)+(Z)
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 236-449-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only