Learn More
Cincinnati Surgical Swann Morton Sterile Carbon Steel Blades
For use in wide variety of medical applications, including Dermaplaning
Supplier:  Cincinnati Surgical 00SM20
 View more versions of this product
Catalog No. 19200233
Only null left
Description
Description
  • 200% visual inspection
  • Packaged with VCI Paper to protect blade from corrosion
  • Strongest/smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended Range
Specifications
Chemical Identifiers
Carbon Steel
Blade
High Surface Finish
Sterile
Specifications
Carbon Steel
# 20
Sterile
Blade
High Surface Finish
Product Suggestions
Product Suggestions