Learn More
Cincinnati Surgical Swann Morton Sterile Carbon Steel Blades
For use in wide variety of medical applications, including Dermaplaning
Supplier: Cincinnati Surgical 00SM15
Description
- 200% visual inspection
- Packaged with VCI Paper to protect blade from corrosion
- Strongest/smallest foil package available
- Narrow grinding angle resulting in increased initial keenness
- High surface finish
- Cellophane overwrap
- Extended Range
Chemical Identifiers
| Carbon Steel | |
| Blade | |
| High Surface Finish | |
| Sterile |
Specifications
| Carbon Steel | |
| # 15 | |
| Sterile |
| Blade | |
| High Surface Finish |