missing translation for 'onlineSavingsMsg'
Learn More

Cinchonidine Sulfate Dihydrate 98.0+%, TCI America™

Catalog No. C034925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C034925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C034925G Supplier TCI America Supplier No. C034925G

Chemical Identifiers

CAS 524-61-8
Molecular Formula C19H28N2O7S
Molecular Weight (g/mol) 428.5
MDL Number MFCD00067565
InChI Key WROWYMULWJEBQS-CNGLPXLGSA-N
PubChem CID 118988797
IUPAC Name (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;sulfuric acid;dihydrate
SMILES C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O.O.O.OS(=O)(=O)O

Specifications

Color White
Boiling Point 240°C
Quantity 25 g
Formula Weight 490.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cinchonidine Sulfate Dihydrate
RTECSNumber GD3050000
TSCA No