CAS RN 524-61-8
IUPAC Name:
(R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol; sulfuric acid
Synonyms:
cinchonidine; sulfuric acid
Molecular Weight (g/mol):
392.47
Molecular Formula:
C19H24N2O5S
InChi Key:
KFHZMXDENPHHOU-YYXOUSRLSA-N
SMILES:
OS(O)(=O)=O.O[C@@H]([C@@H]1C[C@@H]2CCN1C[C@@H]2C=C)C1=C2C=CC=CC2=NC=C1
1
–
1
of
1
results
1
Cinchonidine Sulfate Dihydrate 98.0+%, TCI America™
CAS: 524-61-8 Molecular Formula: C19H28N2O7S Molecular Weight (g/mol): 428.5 MDL Number: MFCD00067565 InChI Key: WROWYMULWJEBQS-CNGLPXLGSA-N PubChem CID: 118988797 IUPAC Name: (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;sulfuric acid;dihydrate SMILES: C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O.O.O.OS(=O)(=O)O