missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cinchonidine, 98.5-101%

Cinchonidine, CAS # 485-71-2, is a naturally occurring alkaloid that may be used in asymmetric synthesis and exhibits anti-malarial activity | CAS: 485-71-2 | C19H22N2O | 294.40 g/mol

Supplier:  Thermo Scientific Chemicals 110331000

 View more versions of this product

Catalog No. AC110331000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cinchonidine is an alkaloid naturally occurring in the bark of Cinchona strubs that has activity against Plasmodium spp
  • Cinchonidine is a diastereoisomer of cinchonine and can be used to promote chirality in asymmetric or enantioselective synthesis of organic compounds

Applications

  • Cinchonidine has been used as a starting material in the synthesis of anti-malarial cinchona alkaloid conjugates
  • Has been shown to potently induce γ-globin mRNA and fetal hemoglobin in erythroid progenitor cells and may be useful in preclinical studies of β-thalassemia
Specifications

Specifications

Cinchonidine
485-71-2
White to Cream
Authentic
Glass bottle
MFCD00006783
23, 420
− 115.00
Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform
OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1
294.40
294.39
Crystalline Powder
98.5-101%
200.0°C to 203.0°C
1% max. (105°C)
98.5-101%
C19H22N2O
100 g
15, 2289
cinchonidine
KMPWYEUPVWOPIM-UHFFFAOYNA-N
[(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
45358337
98.5-101%

Chemical Identifiers

485-71-2
294.40
KMPWYEUPVWOPIM-UHFFFAOYNA-N
45358337
OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1
C19H22N2O
MFCD00006783
cinchonidine
[(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 207-622-3

RTECSNumber : GD2975000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only