Learn More
Invitrogen™ Rat CINC-3 (CXCL2) ELISA Kit
ELISA
Supplier: Invitrogen™ ERCXCL2
Description
The Rat CINC-3 (CXCL2) ELISA quantitates Rt CINC-3 in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt CINC-3. Principle of the method The Rat CINC-3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.Chemical Identifiers
| P30348 | |
| 2 pg/mL | |
| HRP | |
| Chemokine & Receptors | |
| This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 114105 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| Mip-2, Scyb2 | |
| 1 hrs 20 mins | |
| Solid-phase sandwich ELISA |
| 7.37 to 1800 pg/mL | |
| 2 pg/mL to 1800 pg/mL | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 Assays | |
| Colorimetric Microplate Reader | |
| Mip-2,Scyb2 | |
| <10 | |
| HRP | |
| Immunology | |
| 2°C to 8°C | |
| CXCL2 | |
| 4 hrs 45 mins |
Specifications
| P30348 | |
| 2 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 Assays | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 114105 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Immunology | |
| 2°C to 8°C | |
| CXCL2 | |
| 4 hrs 45 mins |
| 7.37 to 1800 pg/mL | |
| 2 pg/mL to 1800 pg/mL | |
| HRP | |
| Chemokine & Receptors | |
| This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha. | |
| Rat | |
| Colorimetric Microplate Reader | |
| Mip-2,Scyb2 | |
| <10 | |
| HRP | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| Mip-2, Scyb2 | |
| 1 hrs 20 mins |