missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Cimetidine USP
Description
Cimetidine USP is a white to off-white, crystalline powder; odorless, or having a slight mercaptan odor.
Specifications
Specifications
| Chemical Name or Material | Cimetidine USP |
| Packaging | Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 51481-61-9 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 252.34 |
| Quantity | 25 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |