missing translation for 'onlineSavingsMsg'
Learn More

Cilostazol 98.0+%, TCI America™

Catalog No. p-7125545
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C25871G 1 g
C25875G 5 g
2 options

Catalog No. C25871G

Supplier: TCI America C25871G

Only null left

Chemical Identifiers

CAS 73963-72-1
Molecular Formula C20H27N5O2
Molecular Weight (g/mol) 369.469
MDL Number MFCD00866780
InChI Key RRGUKTPIGVIEKM-UHFFFAOYSA-N
Synonym cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
PubChem CID 2754
ChEBI CHEBI:31401
IUPAC Name 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4

Specifications

Melting Point 160°C
Color White
Quantity 1 g
Formula Weight 369.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cilostazol
RTECSNumber VC8277500
TSCA No