missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cilostazol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 160°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 369.47 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cilostazol |
Safety and Handling
| RTECSNumber | VC8277500 |
| TSCA | No |