missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chymostatin

Catalog No. AAJ63275LB0
Change view
Click to view available options
Quantity:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63275LB0 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63275LB0 Supplier Thermo Scientific Chemicals Supplier No. J63275LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

A highly specific chymotrypsin inhibitor produced by actinomycetes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

For Use With (Application) Chymotrypsin Inhibitor
Formulation Lyophilized powder
Quantity 5mg
Regulatory Status RUO
Product Type Chymostatin
Recombinant Native
Form Powder

RUO – Research Use Only