missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHSY3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa560777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560777 Supplier Invitrogen™ Supplier No. PA560777
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: LVIILFSRDS GQDSSKHIEL IKGYQNKYPK AEMTLIPMKG EFSRGLGLEM ASAQFDNDTL LLFCDVDLIF R Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

Has both beta-1,3-glucuronic acid and beta-1,4-N-acetylgalactosamine transferase activity. Transfers glucuronic acid (GlcUA) from UDP-GlcUA and N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of the elongating chondroitin polymer. Specific activity is much reduced compared to CHSY1.

Specifications

Antigen CHSY3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHSY3
Gene Accession No. Q70JA7
Gene Alias 4833446K15Rik; AI662215; carbohydrate synthase 2; Chondroitin glucuronyltransferase 3; chondroitin sulfate synthase 3; Chondroitin synthase 2; chondroitin synthase-2; Chsy2; chSy-2; Chsy3; Css3; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II; Kiaa4168; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 3; N-acetylgalactosaminyltransferase 3; RGD1560819
Gene Symbols CHSY3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHSY3. Recombinant protein control fragment (Product #RP-96969).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 337876
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less