missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563163
Description
Immunogen sequence: IISRVSDKLK QIPQALADAN STDPALILAE NASLLSLSEL DSAFSQLQSR L Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
CHST3 is an enzyme which catalyzes the sulfation of chondroitin, a proteoglycan found in the extracellular matrixand most cells which is involved in cell migration and differentiation.Chemical Identifiers
| CHST3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| Chst3 | |
| C6ST; C6ST1; C6ST-1; carbohydrate (chondroitin 6) sulfotransferase 3; carbohydrate (chondroitin 6/keratan) sulfotransferase 3; carbohydrate sulfotransferase 3; chondroitin 6 sulfotransferase 1; chondroitin 6-O-sulfotransferase 1; Chondroitin 6-sulfotransferase; chondroitin 6-sulfotransferase 3; Chst3; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0; GST; Gst0; GST-0; HSD | |
| Chst3 | |
| Primary | |
| Antigen affinity chromatography |
| 0.3 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7LGC8 | |
| 9469 | |
| Recombinant protein fragment of Human CHST3 (Product #RP-100930) | |
| Antibody |
Specifications
| CHST3 | |
| Polyclonal | |
| Unconjugated | |
| Chst3 | |
| C6ST; C6ST1; C6ST-1; carbohydrate (chondroitin 6) sulfotransferase 3; carbohydrate (chondroitin 6/keratan) sulfotransferase 3; carbohydrate sulfotransferase 3; chondroitin 6 sulfotransferase 1; chondroitin 6-O-sulfotransferase 1; Chondroitin 6-sulfotransferase; chondroitin 6-sulfotransferase 3; Chst3; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0; GST; Gst0; GST-0; HSD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9469 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7LGC8 | |
| Chst3 | |
| Recombinant protein fragment of Human CHST3 (Product #RP-100930) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |