missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromone-3-carboxylic Acid 98.0+%, TCI America™
$88.56 - $273.38
Chemical Identifiers
| CAS | 39079-62-4 |
|---|---|
| Molecular Formula | C10H6O4 |
| Molecular Weight (g/mol) | 190.154 |
| MDL Number | MFCD00017338 |
| InChI Key | PCIITXGDSHXTSN-UHFFFAOYSA-N |
| Synonym | chromone-3-carboxylic acid, 4-oxo-4h-chromene-3-carboxylic acid, 4-oxo-4h-1-benzopyran-3-carboxylic acid, maybridge1_005842, 3-carboxychromone, d04tgn, acmc-1ad65, 4-oxo-chromene-3-carboxylic acid, 4h-chromen-4-one-3-carboxylic acid |
| PubChem CID | 181620 |
| IUPAC Name | 4-oxochromene-3-carboxylic acid |
| SMILES | C1=CC=C2C(=C1)C(=O)C(=CO2)C(=O)O |
Chemical Identifiers
| 39079-62-4 | |
| 190.154 | |
| PCIITXGDSHXTSN-UHFFFAOYSA-N | |
| 181620 | |
| C1=CC=C2C(=C1)C(=O)C(=CO2)C(=O)O |
| C10H6O4 | |
| MFCD00017338 | |
| chromone-3-carboxylic acid, 4-oxo-4h-chromene-3-carboxylic acid, 4-oxo-4h-1-benzopyran-3-carboxylic acid, maybridge1_005842, 3-carboxychromone, d04tgn, acmc-1ad65, 4-oxo-chromene-3-carboxylic acid, 4h-chromen-4-one-3-carboxylic acid | |
| 4-oxochromene-3-carboxylic acid |
Specifications
| 39079-62-4 | |
| C10H6O4 | |
| 1 g | |
| PCIITXGDSHXTSN-UHFFFAOYSA-N | |
| 4-oxochromene-3-carboxylic acid | |
| 181620 | |
| ≥98.0% (GC) | |
| Chromone-3-carboxylic Acid |
| 201°C | |
| MFCD00017338 | |
| chromone-3-carboxylic acid, 4-oxo-4h-chromene-3-carboxylic acid, 4-oxo-4h-1-benzopyran-3-carboxylic acid, maybridge1_005842, 3-carboxychromone, d04tgn, acmc-1ad65, 4-oxo-chromene-3-carboxylic acid, 4h-chromen-4-one-3-carboxylic acid | |
| C1=CC=C2C(=C1)C(=O)C(=CO2)C(=O)O | |
| 190.154 | |
| 190.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No