accessibility menu, dialog, popup

UserName

CAS RN 39079-62-4

IUPAC Name: 4-oxo-4H-chromene-3-carboxylate
Synonyms: 4-oxochromene-3-carboxylate
Molecular Weight (g/mol): 189.15
Molecular Formula: C10H5O4
InChi Key: PCIITXGDSHXTSN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=COC2=CC=CC=C2C1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)