missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. C23675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23675G 5 g
C23671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23675G Supplier TCI America Supplier No. C23675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39079-62-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00017338
InChI Key PCIITXGDSHXTSN-UHFFFAOYSA-N
Synonym chromone-3-carboxylic acid, 4-oxo-4h-chromene-3-carboxylic acid, 4-oxo-4h-1-benzopyran-3-carboxylic acid, maybridge1_005842, 3-carboxychromone, d04tgn, acmc-1ad65, 4-oxo-chromene-3-carboxylic acid, 4h-chromen-4-one-3-carboxylic acid
PubChem CID 181620
IUPAC Name 4-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C(=O)O

Specifications

Melting Point 201°C
Quantity 5 g
Formula Weight 190.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chromone-3-carboxylic Acid