missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carboxylic acid, 98%

Catalog No. AAA1442203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1442203 1 g
AAA1442206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1442203 Supplier Thermo Scientific Chemicals Supplier No. A1442203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39079-62-4 | C10H6O4 | 190.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39079-62-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00017338
InChI Key PCIITXGDSHXTSN-UHFFFAOYSA-N
Synonym chromone-3-carboxylic acid, 4-oxo-4h-chromene-3-carboxylic acid, 4-oxo-4h-1-benzopyran-3-carboxylic acid, maybridge1_005842, 3-carboxychromone, d04tgn, acmc-1ad65, 4-oxo-chromene-3-carboxylic acid, 4h-chromen-4-one-3-carboxylic acid
PubChem CID 181620
IUPAC Name 4-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C(=O)O

Specifications

Melting Point 198°C to 203°C
Quantity 1 g
Beilstein 1074111
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material Chromone-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only