missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carbonitrile 98.0+%, TCI America™

Catalog No. C30755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30755G Supplier TCI America Supplier No. C30755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50743-17-4
Molecular Formula C10H5NO2
Molecular Weight (g/mol) 171.16
MDL Number MFCD00052604
InChI Key SFWNPLLGXKJESA-UHFFFAOYSA-N
Synonym 3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile
PubChem CID 521298
IUPAC Name 4-oxo-4H-chromene-3-carbonitrile
SMILES O=C1C(=COC2=CC=CC=C12)C#N

Specifications

Melting Point 177°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 171.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chromone-3-carbonitrile