accessibility menu, dialog, popup

UserName

CAS RN 50743-17-4

IUPAC Name: 4-oxo-4H-chromene-3-carbonitrile
Synonyms: 4-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 171.16
Molecular Formula: C10H5NO2
InChi Key: SFWNPLLGXKJESA-UHFFFAOYSA-N
SMILES: O=C1C(=COC2=CC=CC=C12)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results