missing translation for 'onlineSavingsMsg'
Learn More

Chromone-2-carboxylic Acid 98.0+%, TCI America™

$316.89 - $1127.75

Chemical Identifiers

CAS 4940-39-0
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00006838
InChI Key RVMGXWBCQGAWBR-UHFFFAOYSA-N
Synonym chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
PubChem CID 2741
IUPAC Name 4-oxochromene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O02905G
View Documents
TCI America
O02905G
5 g
Each for $316.89
Only null left
 
O029025G
View Documents
TCI America
O029025G
25 g
Each for $1,127.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4940-39-0
190.154
RVMGXWBCQGAWBR-UHFFFAOYSA-N
2741
C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O
C10H6O4
MFCD00006838
chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
4-oxochromene-2-carboxylic acid

Specifications

4940-39-0
MFCD00006838
chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O
190.154
190.15
Crystalline Powder
C10H6O4
5 g
RVMGXWBCQGAWBR-UHFFFAOYSA-N
4-oxochromene-2-carboxylic acid
2741
≥98.0% (HPLC,T)
Chromone-2-carboxylic Acid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ2476000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'