missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromone-2-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America O029025G
Specifications
| Chromone-2-carboxylic Acid | |
| C10H6O4 | |
| 25 g | |
| RVMGXWBCQGAWBR-UHFFFAOYSA-N | |
| 4-oxochromene-2-carboxylic acid | |
| 2741 | |
| ≥98.0% (HPLC,T) |
| 4940-39-0 | |
| MFCD00006838 | |
| chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french | |
| C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O | |
| 190.154 | |
| 190.15 | |
| Crystalline Powder |
Chemical Identifiers
| 4940-39-0 | |
| 190.154 | |
| RVMGXWBCQGAWBR-UHFFFAOYSA-N | |
| 2741 | |
| C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O |
| C10H6O4 | |
| MFCD00006838 | |
| chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french | |
| 4-oxochromene-2-carboxylic acid |
Safety and Handling
RTECSNumber : DJ2476000
TSCA : No