missing translation for 'onlineSavingsMsg'
Learn More

Chromium VI - Soil, MilliporeSigma™ Supelco™

Catalog No. 111001647
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001647 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001647 Supplier Merck Emd Millipore Supplier No. CRM04130G

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental

Specifications

CAS 14808-60-7
Type Certified Reference Material
Quantity 30 g
Grade Certified Reference Material