missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium ICP Standard, 10,000 ppm Cr6+ in 5% HCl, Ricca Chemical

Catalog No. p-3340157
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PCR610KH50 Natural Poly Bottle
PCR610KH100 Natural LDPE Bottle
PCR610KH500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PCR610KH50 Supplier Ricca Chemical Company Supplier No. PCR610KH50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7789-09-5 , 7647-01-0 , 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Packaging Natural Poly Bottle
CAS Min % 90.0, 5.34, 2.19
CAS Max % 94.0, 5.56, 2.28
Concentration or Composition (by Analyte or Components) 1mL = 10mg Cr (10, 000ppm Cr)(NH4)2Cr2O7 in 5% HCl
Chemical Name or Material Chromium(VI) ICP Standard
ShelfLife 18 months