missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Trioxide, ACS Reagent (Chromic Acid), Reagents
Chromium Trioxide, ACS Reagent (Chromic Acid), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C117950025KG
Specifications
| 250°C | |
| 196°C | |
| 1333-82-0 | |
| 2.5 kg | |
| MFCD00010952 | |
| WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
| trioxochromium | |
| ACS | |
| 2.7 |
| Chromium Trioxide, ACS Reagent (Chromic Acid) | |
| Dark red-Black | |
| Solid | |
| CrO3 | |
| Chromic Acid | |
| O=[Cr](=O)=O | |
| 99.99 | |
| Glass Bottle |
Chemical Identifiers
| 1333-82-0 | |
| 99.99 | |
| WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
| trioxochromium |
| CrO3 | |
| MFCD00010952 | |
| Chromic Acid | |
| O=[Cr](=O)=O |