missing translation for 'onlineSavingsMsg'
Learn More

Chromium Trioxide, ACS Reagent (Chromic Acid), Reagents

Catalog No. p-200021191
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338044 100 g
01338043 2.5 kg
01338042 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338044 Supplier Labchem Inc Supplier No. C1179500100C2
Only null left

Chromium Trioxide, ACS Reagent (Chromic Acid), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym Chromic Acid
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Boiling Point 250°C
Melting Point 196°C
Color Dark red-Black
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.7
Chemical Name or Material Chromium Trioxide, ACS Reagent (Chromic Acid)