Learn More
Chromium silicide, 99+% (metals basis)
CAS: 12018-09-6 | CrSi2 | 108.166 g/mol
$139.63 - $515.47
Chemical Identifiers
| CAS | 12018-09-6 |
|---|---|
| Molecular Formula | CrSi2 |
| Molecular Weight (g/mol) | 108.166 |
| MDL Number | MFCD00168070 |
| InChI Key | CHXGWONBPAADHP-UHFFFAOYSA-N |
| Synonym | chromsilicid, chromium silicide, bis $l^ 3 |
| PubChem CID | 45051936 |
| IUPAC Name | bis($l^{3}-silanylidyne)chromium |
| SMILES | [Si]#[Cr]#[Si] |
Description
Chromium silicide is applied in production of ceramic materials. It can also produce an oxidation-resistant surface in a low pressure plasma spraying.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12018-09-6 | |
| 108.166 | |
| CHXGWONBPAADHP-UHFFFAOYSA-N | |
| 45051936 | |
| [Si]#[Cr]#[Si] |
| CrSi2 | |
| MFCD00168070 | |
| chromsilicid, chromium silicide, bis $l^ 3 | |
| bis($l^{3}-silanylidyne)chromium |
Specifications
| 1490°C | |
| Gray | |
| 50 g | |
| CrSi2 | |
| chromsilicid, chromium silicide, bis $l^ 3 | |
| CHXGWONBPAADHP-UHFFFAOYSA-N | |
| bis($l^{3}-silanylidyne)chromium | |
| 45051936 | |
| Odorless | |
| Gray powder |
| 12018-09-6 | |
| Powder | |
| >95% | |
| MFCD00168070 | |
| Insoluble in water. | |
| [Si]#[Cr]#[Si] | |
| 108.166 | |
| 108.17 | |
| Moisture sensitive | |
| Chromium silicide |
Safety and Handling
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 234-633-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only