accessibility menu, dialog, popup

UserName

SiCrSi

CAS RN 12018-09-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 108.17
Molecular Formula: CrSi2
InChi Key: CHXGWONBPAADHP-UHFFFAOYSA-N
SMILES: [Si]#[Cr]#[Si]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results