Learn More
Chromium powder, -100 mesh, 99% (metals basis)
Apparent density 2.0-3.0g/cm3 | CAS: 7440-47-3 | Cr | 52.00 g/mol
Supplier: Thermo Scientific Chemicals 01014722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gray | |
| 7440-47-3 | |
| 100 g | |
| Odorless | |
| 99% (Metals basis) | |
| MFCD00010944 | |
| VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
| chromium | |
| 23976 |
| Chromium | |
| Gray powder | |
| -100 Mesh | |
| Powder | |
| Cr | |
| chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
| [Cr] | |
| 52.00 | |
| CHEBI:28073 |
Chemical Identifiers
| 7440-47-3 | |
| 52.00 | |
| VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
| 23976 | |
| chromium |
| Cr | |
| MFCD00010944 | |
| chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
| CHEBI:28073 | |
| [Cr] |
Safety and Handling
Wear suitable protective clothing and gloves.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-157-5
RTECSNumber : GB4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only