missing translation for 'onlineSavingsMsg'
Learn More

Chromium powder, -100 mesh, 99% (metals basis)

Catalog No. p-4781854
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1014709 10 g
AA1014722 100 g
AA1014736 500 g
AA10147A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1014709 Supplier Thermo Scientific Chemicals Supplier No. 01014709
Only null left

Apparent density 2.0-3.0g/cm3 | CAS: 7440-47-3 | Cr | 52.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Color Gray
Appearance Gray powder
Quantity 10 g
Mesh Size -100 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99% (Metals basis)
Chemical Name or Material Chromium
Hazard Category H228
Hazard Statement Wear suitable protective clothing and gloves.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-157-5
RTECSNumber GB4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only