missing translation for 'onlineSavingsMsg'
Learn More

Chromium nitride

CAS: 12053-27-9 | CrN | 66.00 g/mol

$140.86 - $379.53

Chemical Identifiers

CAS 12053-27-9
Molecular Formula CrN
Molecular Weight (g/mol) 66.00
MDL Number MFCD00016048
InChI Key YTZQKGWLQOLFNF-UHFFFAOYSA-N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1214914
View Documents
Thermo Scientific Chemicals
01214914
25 g
Each for $140.86
Only null left
 
AA1214922
View Documents
Thermo Scientific Chemicals
01214922
100 g
Each for $379.53
Only null left
 
Description

Description

Chromium nitride is used for stainless steel, corrosion resistance steel, alloy steel and such special steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12053-27-9
66.00
YTZQKGWLQOLFNF-UHFFFAOYSA-N
CrN
MFCD00016048

Specifications

12053-27-9
25 g
MFCD00016048
YTZQKGWLQOLFNF-UHFFFAOYSA-N
Chromium nitride
Powder
CrN
Insoluble in water.
66.00
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

EINECSNumber : 235-002-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only