accessibility menu, dialog, popup

UserName

CAS RN 12053-27-9

IUPAC Name: chromium(3+) azanetriide
Synonyms: chromium(3+) azanetriide
Molecular Weight (g/mol): 66
Molecular Formula: CrN
InChi Key: YTZQKGWLQOLFNF-UHFFFAOYSA-N
SMILES: [N-3].[Cr+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results