missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) Pyridine-2-carboxylate 96.0+%, TCI America™
$109.98 - $372.81
Chemical Identifiers
| CAS | 14639-25-9 |
|---|---|
| Molecular Formula | C18H12CrN3O6 |
| Molecular Weight (g/mol) | 418.305 |
| MDL Number | MFCD00068715 |
| InChI Key | CBDQOLKNTOMMTL-UHFFFAOYSA-K |
| Synonym | chromium picolinate, chromium iii pyridine-2-carboxylate, chromium iii picolinate, chromium tripicolinate, picolinic acid chromium iii salt, chromium, tris picolinato, unii-s71t8b8z6p, chromium iii trispicolinate, picolinic acid, chromium salt, chromium 2-pyridinecarboxylate |
| PubChem CID | 56840998 |
| ChEBI | CHEBI:50369 |
| IUPAC Name | chromium(3+);pyridine-2-carboxylate |
| SMILES | C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cr+3] |
Chemical Identifiers
| 14639-25-9 | |
| 418.305 | |
| CBDQOLKNTOMMTL-UHFFFAOYSA-K | |
| 56840998 | |
| chromium(3+);pyridine-2-carboxylate |
| C18H12CrN3O6 | |
| MFCD00068715 | |
| chromium picolinate, chromium iii pyridine-2-carboxylate, chromium iii picolinate, chromium tripicolinate, picolinic acid chromium iii salt, chromium, tris picolinato, unii-s71t8b8z6p, chromium iii trispicolinate, picolinic acid, chromium salt, chromium 2-pyridinecarboxylate | |
| CHEBI:50369 | |
| C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cr+3] |
Specifications
| 14639-25-9 | |
| C18H12CrN3O6 | |
| 5 g | |
| CBDQOLKNTOMMTL-UHFFFAOYSA-K | |
| chromium(3+);pyridine-2-carboxylate | |
| 56840998 | |
| 418.31 | |
| Crystalline Powder |
| Purple-Red | |
| MFCD00068715 | |
| chromium picolinate, chromium iii pyridine-2-carboxylate, chromium iii picolinate, chromium tripicolinate, picolinic acid chromium iii salt, chromium, tris picolinato, unii-s71t8b8z6p, chromium iii trispicolinate, picolinic acid, chromium salt, chromium 2-pyridinecarboxylate | |
| C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cr+3] | |
| 418.305 | |
| CHEBI:50369 | |
| ≥96.0% (T) | |
| Chromium(III) Pyridine-2-carboxylate |
Safety and Handling
RTECSNumber : US5647790
TSCA : No