accessibility menu, dialog, popup

UserName

CAS RN 14639-25-9

CrOONOONOON

CAS RN 14639-25-9

IUPAC Name: chromium(3+) tris(pyridine-2-carboxylate)
Synonyms: chromium(III) picolinate
Molecular Weight (g/mol): 418.31
Molecular Formula: C18H12CrN3O6
InChi Key: CBDQOLKNTOMMTL-UHFFFAOYSA-K
SMILES: [Cr+3].[O-]C(=O)C1=CC=CC=N1.[O-]C(=O)C1=CC=CC=N1.[O-]C(=O)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results