missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, 90%

Catalog No. AC223810050
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223810050 5 g Glass bottle
AC223810100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223810050 Supplier Thermo Scientific Chemicals Supplier No. 223810050

CAS: 10049-05-5 | Cl2Cr | 122.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 10049-05-5
Molecular Formula Cl2Cr
Molecular Weight (g/mol) 122.9
MDL Number MFCD00010947
Synonym Chromous chloride

Specifications

Melting Point 824.0°C
Color Gray to Green
Physical Form Powder
Quantity 5 g
Assay Percent Range 88% min. (Argentometry)
Linear Formula CrCl2
Fieser 01,149; 02,76; 03,60; 04,144; 07,73; 09,511; 12,136; 13,84; 14,94; 16,93; 17,84
Merck Index 15, 2248
Solubility Information Solubility in water: may decompose. Other solubilities: insoluble in alcohol and ether
Formula Weight 122.9
Percent Purity 90%
Packaging Glass bottle
Chemical Name or Material Chromium(II) chloride, 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical
EINECSNumber 233-163-3
RTECSNumber GB5250000
TSCA TSCA

RUO – Research Use Only