CAS RN 10049-05-5
IUPAC Name:
λ²-chromium(2+) dichloride
Synonyms:
chromium(II) chloride
Molecular Weight (g/mol):
122.9
Molecular Formula:
Cl2Cr
InChi Key:
XBWRJSSJWDOUSJ-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].[Cr++]
1
–
3
of
3
results
1
Chromium(II) chloride, 99.9%, (trace metal basis)
CAS: 10049-05-5 Molecular Formula: Cl2Cr Molecular Weight (g/mol): 122.9 MDL Number: MFCD00010947 Synonym: Chromous chloride
2
Chromium(II) chloride, 90%
CAS: 10049-05-5 Molecular Formula: Cl2Cr Molecular Weight (g/mol): 122.9 MDL Number: MFCD00010947 Synonym: Chromous chloride
3
Chromium(II) chloride, anhydrous, 97%
CAS: 10049-05-5 Molecular Formula: CrCl2 MDL Number: MFCD00010947 Synonym: Chromous chloride