missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromic Chloride, Hexahydrate, USP, 98-101%, Spectrum™ Chemical
C1344, 10060-12-5, CrCl3·6H2O
Supplier: Spectrum Chemical Mfg Cor C134412KGBL
Description
Spectrum™ Chemical Chromic Chloride, Hexahydrate, USP is intended for use as an additive to intravenous solutions for total parenteral nutrition. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 10060-12-5 | |
| Cl3CrH12O6 | |
| 12 kg | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] | |
| 266.44 | |
| USP | |
| Poly Pail |
| 100% | |
| MFCD00149660 | |
| LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
| chromium(3+) hexahydrate trichloride | |
| 98 to 101% | |
| 0.01% |
Chemical Identifiers
| 10060-12-5 | |
| 266.44 | |
| LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
| Cl3CrH12O6 | |
| MFCD00149660 | |
| chromium(3+) hexahydrate trichloride |