missing translation for 'onlineSavingsMsg'
Learn More

Chromic Chloride, Hexahydrate, USP, 98-101%, Spectrum™ Chemical

Catalog No. p-7157153
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601686 125 g Amber Glass Bottle
18601687 500 g Amber Glass Bottle
18601689 2.5 kg Poly Pail
18601688 12 kg Poly Pail
18601690 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601686 Supplier Spectrum Chemical Mfg Cor Supplier No. C1344125GM
Only null left

C1344, 10060-12-5, CrCl3·6H2O

Spectrum™ Chemical Chromic Chloride, Hexahydrate, USP is intended for use as an additive to intravenous solutions for total parenteral nutrition. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 98 to 101%
Grade USP
Insoluble Matter 0.01%
Packaging Amber Glass Bottle