missing translation for 'onlineSavingsMsg'
Learn More

Chromic Acid, 30% (w/w), Reagents

Catalog No. 01335570
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335570 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335570 Supplier Labchem Inc Supplier No. CC2410001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chromic Acid, 30% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1333-82-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chromium trioxide solution
IUPAC Name water
SMILES O

Specifications

Color Orange/Red
Boiling Point 106°C
Packaging Poly Bottle
Specific Gravity 1.2326
Quantity 1 L
Physical Form Liquid
Percent Purity 30%
Grade Reagent
Chemical Name or Material Chromic Acid, 30% (w/w)