missing translation for 'onlineSavingsMsg'
Learn More

Chromic Acid, 10% w/v, Reagents

Catalog No. 01335673
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335673 1 L
01335672 4 L
01335671 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335673 Supplier Labchem Inc Supplier No. CC1011001A
Only null left

Chromic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1333-82-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chromium trioxide solution
IUPAC Name water
SMILES O

Specifications

Color Orange
Boiling Point 101°C
Packaging Poly Bottle
Specific Gravity 1.071
Quantity 1 L
Physical Form Liquid
Percent Purity 10% w/v
Grade Reagent
Chemical Name or Material Chromic Acid, 10% w/v