missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromic Acid, 10% w/v, Reagents
Chromic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC101100500A
Specifications
| Chromic Acid, 10% w/v | |
| 7732-18-5,1333-82-0 | |
| Poly Bottle | |
| 500 mL | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 10% w/v |
| Orange | |
| 101°C | |
| 1.071 | |
| H2O | |
| Chromium trioxide solution | |
| O | |
| 18.02 | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Chromium trioxide solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |